Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027839.2 | NP_082115.2 | 4 - 5 | 772 | 70 | |
GenBank mRNA | AK018613.1 | - | 4 - 5 | 769 | 70 | |
AK033581.2 | - | 4 - 5 | 773 | 70 | ||
BC049361.1 | - | 4 - 5 | 753 | 6476373 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |