Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027839.2 | NP_082115.2 | 10 - 11 | 1643 | 131 | |
GenBank mRNA | AK033581.2 | - | 10 - 11 | 1644 | 131 | |
BC049361.1 | - | 10 - 11 | 1624 | 6476373 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |