Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282054.1 | NP_001268983.1 | 15 - 16 | 2040 | 73 | |
NM_031870.3 | NP_114076.1 | 14 - 15 | 2131 | 73 | ||
GenBank mRNA | AF178957.1 | - | 14 - 15 | 1835 | 73 | |
AF298655.1 | - | 14 - 15 | 2131 | 73 | ||
AK029837.1 | - | 15 - 16 | 2041 | 73 | ||
AK161474.1 | - | 14 - 15 | 1881 | 73 | ||
AY351585.1 | - | 14 - 15 | 2085 | 73 | ||
AY351587.1 | - | 3 - 4 | 1082 | 73 | ||
AY351590.1 | - | 6 - 7 | 1547 | 73 | ||
BC138163.1 | - | 14 - 15 | 2022 | 8861182 | 73 | |
BC145838.1 | - | 14 - 15 | 2022 | 40131141 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |