Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026582.4 | NP_080858.3 | 8 - 9 | 1385 | 144 | |
NR_037590.1 | 9 - 10 | 1466 | 144 | |||
GenBank mRNA | AK019886.1 | - | 9 - 10 | 1424 | 144 | |
AK029053.1 | - | 8 - 9 | 1344 | 144 | ||
AK029104.1 | - | 8 - 9 | 1342 | 144 | ||
AK044810.1 | - | 8 - 9 | 1344 | 144 | ||
AK049842.1 | - | 8 - 9 | 1342 | 144 | ||
AK077407.1 | - | 8 - 9 | 1343 | 144 | ||
AK128912.1 | - | 8 - 9 | 1341 | 144 | ||
AK159207.1 | - | 8 - 9 | 1342 | 144 | ||
BC018381.1 | - | 7 - 8 | 969 | 4237619 | 144 | |
BC075615.1 | - | 8 - 9 | 1385 | 5703610 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |