Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100015350.mm.1 |
NM_001134829.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015350.mm.1 |
NM_001134829.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015350.mm.1 |
TC0100001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015353.mm.1 |
NM_001134829.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015353.mm.1 |
NM_001134829.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015353.mm.1 |
NM_172266.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015353.mm.1 |
NM_172266.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015353.mm.1 |
TC0100001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015348.mm.1 |
NM_001134829.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015348.mm.1 |
NM_001134829.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015348.mm.1 |
TC0100001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015349.mm.1 |
NM_001134829.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015349.mm.1 |
NM_001134829.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015349.mm.1 |
TC0100001855.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015352.mm.1 |
NM_001134829.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015352.mm.1 |
NM_001134829.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015352.mm.1 |
NM_172266.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015352.mm.1 |
NM_172266.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015352.mm.1 |
TC0100001855.mm.1 |
MTA TC |