Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008486.2 | NP_032512.2 | 7 - 8 | 1534 | 69 | |
GenBank mRNA | AK143746.1 | - | 7 - 8 | 1477 | 69 | |
AK171427.1 | - | 8 - 9 | 1896 | 69 | ||
BC005431.1 | - | 7 - 8 | 1500 | 3593307 | 69 | |
BC017011.1 | - | 7 - 8 | 1558 | 3590117 | 69 | |
BC040792.1 | - | 7 - 8 | 1558 | 3590117 | 69 | |
U77083.1 | - | 7 - 8 | 1466 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |