Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017474.2 | NP_059502.1 | 7 - 8 | 1219 | 91 | |
GenBank mRNA | AB017156.1 | - | 7 - 8 | 1202 | 91 | |
AK007466.1 | - | 7 - 8 | 1216 | 91 | ||
AK008659.2 | - | 7 - 8 | 1216 | 91 | ||
AK033560.1 | - | 1220 | 91 | |||
AK136476.1 | - | 7 - 8 | 1216 | 91 | ||
BC028343.1 | - | 7 - 8 | 1209 | 4217548 | 91 | |
BC116318.2 | - | 7 - 8 | 1202 | 40095991 | 91 | |
BC116319.2 | - | 7 - 8 | 1202 | 40095992 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |