Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400016039.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016039.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016039.mm.1 |
TC1400002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016055.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016055.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016055.mm.1 |
TC1400002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016051.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016051.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016051.mm.1 |
TC1400002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016052.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016052.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016052.mm.1 |
TC1400002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016054.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016054.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016054.mm.1 |
TC1400002864.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400016053.mm.1 |
NM_028780.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016053.mm.1 |
NM_028780.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400016053.mm.1 |
TC1400002864.mm.1 |
MTA TC |