Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010209.2 | NP_034339.2 | 3 - 4 | 427 | 91 | |
GenBank mRNA | AK002379.1 | - | 3 - 4 | 379 | 91 | |
AK147000.1 | - | 3 - 4 | 421 | 91 | ||
BC006048.1 | - | 3 - 4 | 370 | 3590691 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |