Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddit4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030143.4 | NP_084419.2 | 1 - 2 | 213 | 68 | |
GenBank mRNA | AK006617.1 | - | 1 - 2 | 212 | 68 | |
AK018944.1 | - | 1 - 2 | 215 | 68 | ||
AK053333.1 | - | 1 - 2 | 151 | 68 | ||
AK075693.1 | - | 1 - 2 | 102 | 68 | ||
BC038131.1 | - | 1 - 2 | 112 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |