Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnaseh2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026001.2 | NP_080277.1 | 9 - 10 | 740 | 67 | |
GenBank mRNA | AK011914.1 | - | 9 - 10 | 779 | 67 | |
AK082752.1 | - | 9 - 10 | 741 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |