Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019649.2 | NP_062623.2 | 1 - 2 | 100 | 74 | |
GenBank mRNA | AK148080.1 | - | 1 - 2 | 82 | 74 | |
AK152410.1 | - | 1 - 2 | 90 | 74 | ||
AK152809.1 | - | 1 - 2 | 90 | 74 | ||
AK165742.1 | - | 1 - 2 | 100 | 74 | ||
AK171901.1 | - | 1 - 2 | 91 | 74 | ||
BC022172.1 | - | 1 - 2 | 91 | 4193279 | 74 | |
BY288640.1 | - | 1 - 2 | 104 | 74 | ||
D67067.1 | - | 1 - 2 | 87 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |