Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qpctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026111.3 | NP_080387.2 | 3 - 4 | 652 | 70 | |
GenBank mRNA | AK007701.1 | - | 3 - 4 | 680 | 70 | |
AK035587.1 | - | 3 - 4 | 652 | 70 | ||
AK045687.1 | - | 3 - 4 | 656 | 70 | ||
AK148755.1 | - | 1 - 2 | 664 | 70 | ||
BC058181.1 | - | 3 - 4 | 675 | 5325516 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |