Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009794.3 | NP_033924.2 | 2 - 3 | 438 | 91 | |
GenBank mRNA | AF015038.1 | - | 2 - 3 | 384 | 91 | |
AK052141.1 | - | 2 - 3 | 413 | 91 | ||
AK150695.1 | - | 2 - 3 | 412 | 91 | ||
AK152094.1 | - | 2 - 3 | 412 | 91 | ||
AK152790.1 | - | 2 - 3 | 412 | 91 | ||
AK152994.1 | - | 2 - 3 | 412 | 91 | ||
AK154855.1 | - | 2 - 3 | 452 | 91 | ||
BC054726.1 | - | 2 - 3 | 438 | 5703563 | 91 | |
D38117.1 | - | 2 - 3 | 479 | 91 | ||
Y10139.1 | - | 2 - 3 | 320 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |