Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163514.1 | NP_001156986.1 | 1 - 2 | 246 | 117 | |
GenBank mRNA | AB032171.1 | - | 1 - 2 | 234 | 117 | |
AF200973.1 | - | 1 - 2 | 211 | 117 | ||
AK019370.1 | - | 1 - 2 | 177 | 117 | ||
BY251302.1 | - | 1 - 2 | 246 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |