Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007934.3 | NP_031960.1 | 16 - 17 | 2570 | 64 | |
GenBank mRNA | AK031439.1 | - | 15 - 16 | 1602 | 64 | |
AK128975.1 | - | 16 - 17 | 2527 | 64 | ||
AK146808.1 | - | 16 - 17 | 2542 | 64 | ||
BC127060.1 | - | 16 - 17 | 2434 | 40109863 | 64 | |
DQ000292.1 | - | 16 - 17 | 2310 | 64 | ||
M29961.1 | - | 16 - 17 | 2389 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |