Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcorl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178782.4 | NP_848897.3 | 9 - 10 | 4548 | 63 | |
GenBank mRNA | AK054259.1 | - | 5 - 6 | 1082 | 63 | |
AK081004.1 | - | 5 - 6 | 3116 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |