Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aifm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290364.1 | NP_001277293.1 | 4 - 5 | 686 | 75 | |
NM_012019.3 | NP_036149.1 | 4 - 5 | 698 | 75 | ||
GenBank mRNA | AF100927.1 | - | 4 - 5 | 524 | 75 | |
AK133764.1 | - | 4 - 5 | 538 | 75 | ||
BC003292.1 | - | 4 - 5 | 561 | 3500037 | 75 | |
BC024882.1 | - | 4 - 5 | 1182 | 75 | ||
DQ016499.1 | - | 4 - 5 | 561 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |