Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs6st2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077202.2 | NP_001070670.1 | 2 - 3 | 948 | 73 | |
NM_001290468.1 | NP_001277397.1 | 1 - 2 | 589 | 73 | ||
GenBank mRNA | AB024565.1 | - | 1 - 2 | 511 | 73 | |
AK043774.1 | - | 1 - 2 | 72 | 73 | ||
AK052348.1 | - | 2 - 3 | 728 | 73 | ||
AK083281.1 | - | 1 - 2 | 576 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |