Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020010.2 | NP_064394.2 | 7 - 8 | 1458 | 98 | |
GenBank mRNA | AF166266.1 | - | 7 - 8 | 1210 | 98 | |
AK028355.1 | - | 7 - 8 | 1151 | 98 | ||
AK028815.1 | - | 7 - 8 | 1215 | 98 | ||
AK031059.1 | - | 7 - 8 | 1191 | 98 | ||
AK075764.1 | - | 7 - 8 | 1190 | 98 | ||
AK076983.1 | - | 7 - 8 | 1191 | 98 | ||
BC031813.1 | - | 7 - 8 | 1185 | 4193738 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |