Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003909.4 | NP_001003909.2 | 6 - 7 | 1610 | 115 | |
NM_001289527.1 | NP_001276456.1 | 6 - 7 | 1610 | 115 | ||
NM_001289528.1 | NP_001276457.1 | 5 - 6 | 1492 | 115 | ||
NM_001289529.1 | NP_001276458.1 | 5 - 6 | 1082 | 115 | ||
GenBank mRNA | AK129343.1 | - | 6 - 7 | 1411 | 115 | |
BC020475.1 | - | 6 - 7 | 1144 | 115 | ||
BC089031.1 | - | 5 - 6 | 1061 | 115 | ||
BC145510.1 | - | 5 - 6 | 1243 | 115 | ||
BC145511.1 | - | 6 - 7 | 1361 | 9054074 | 115 | |
BC150792.1 | - | 6 - 7 | 1361 | 9087703 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |