Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018811.6 | NP_061281.3 | 3 - 4 | 371 | 79 | |
GenBank mRNA | AF201730.1 | - | 3 - 4 | 360 | 79 | |
AF546701.1 | - | 3 - 4 | 372 | 79 | ||
AK008690.1 | - | 3 - 4 | 381 | 79 | ||
AK139526.1 | - | 3 - 4 | 380 | 79 | ||
AK153642.1 | - | 3 - 4 | 418 | 79 | ||
BC027098.1 | - | 3 - 4 | 341 | 5356667 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |