Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018811.6 | NP_061281.3 | 7 - 8 | 997 | 70 | |
GenBank mRNA | AF201730.1 | - | 7 - 8 | 986 | 70 | |
AF546701.1 | - | 7 - 8 | 998 | 70 | ||
AK008690.1 | - | 7 - 8 | 1007 | 70 | ||
AK016296.1 | - | 5 - 6 | 658 | 70 | ||
AK139526.1 | - | 7 - 8 | 1006 | 70 | ||
AK153642.1 | - | 7 - 8 | 1044 | 70 | ||
BC027098.1 | - | 7 - 8 | 967 | 5356667 | 70 | |
BC089477.1 | - | 7 - 8 | 977 | 6741521 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |