Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00013987.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013987.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013987.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013982.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013982.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013982.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013986.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013986.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013986.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013983.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013983.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013983.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013980.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013980.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013980.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013985.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013985.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013985.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013979.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013979.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013979.mm.1 |
TC0X00002187.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013981.mm.1 |
NM_001190332.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013981.mm.1 |
NM_001190332.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013981.mm.1 |
TC0X00002187.mm.1 |
MTA TC |