Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016694.3 | NP_057903.1 | 9 - 10 | 1174 | 76 | |
GenBank mRNA | AB019558.1 | - | 9 - 10 | 1145 | 76 | |
AF250293.1 | - | 9 - 10 | 1174 | 76 | ||
BC113204.1 | - | 9 - 10 | 1157 | 40047891 | 76 | |
FJ555554.1 | - | 7 - 8 | 927 | 76 | ||
FJ555555.1 | - | 4 - 5 | 585 | 76 | ||
FJ555556.1 | - | 8 - 9 | 907 | 76 | ||
FJ555557.1 | - | 8 - 9 | 1032 | 76 | ||
GU272056.1 | - | 3 - 4 | 448 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |