Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031388.2 | NP_113565.2 | 6 - 7 | 533 | 63 | |
GenBank mRNA | AF285570.1 | - | 3 - 4 | 129 | 63 | |
AK082829.1 | - | 6 - 7 | 460 | 63 | ||
AK133417.1 | - | 6 - 7 | 537 | 63 | ||
BC125454.1 | - | 1 - 2 | 32 | 40129969 | 63 | |
BC132154.1 | - | 1 - 2 | 32 | 40130431 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |