Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159850.1 | NP_001153322.1 | 2 - 3 | 618 | 102 | |
NM_001281847.1 | NP_001268776.1 | 2 - 3 | 509 | 102 | ||
NM_010607.3 | NP_034737.2 | 2 - 3 | 714 | 102 | ||
GenBank mRNA | AK138427.1 | - | 2 - 3 | 390 | 102 | |
AK140716.1 | - | 2 - 3 | 389 | 102 | ||
AY736359.1 | - | 2 - 3 | 353 | 102 | ||
BC062094.1 | - | 2 - 3 | 332 | 6848897 | 102 | |
U73488.2 | - | 3 - 4 | 791 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |