Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190717.1 | NP_001177646.1 | 10 - 11 | 1121 | 131 | |
NM_013726.3 | NP_038754.1 | 10 - 11 | 1124 | 131 | ||
GenBank mRNA | AJ003132.1 | - | 10 - 11 | 1014 | 131 | |
AK132403.1 | - | 10 - 11 | 1086 | 131 | ||
AK163408.1 | - | 8 - 9 | 700 | 131 | ||
BC138709.1 | - | 10 - 11 | 942 | 8861731 | 131 | |
BC138710.1 | - | 10 - 11 | 942 | 8861732 | 131 | |
BC145392.1 | - | 10 - 11 | 939 | 9053950 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |