Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023733.3 | NP_076222.1 | 1 - 2 | 43 | 63 | |
GenBank mRNA | AK004567.2 | - | 1 - 2 | 44 | 63 | |
BC006593.1 | - | 1 - 2 | 31 | 3156501 | 63 | |
BC012308.1 | - | 1 - 2 | 58 | 3500167 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |