Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023733.3 | NP_076222.1 | 2 - 3 | 132 | 137 | |
GenBank mRNA | AK004567.2 | - | 2 - 3 | 133 | 137 | |
AK037973.1 | - | 1 - 2 | 584 | 137 | ||
BC006593.1 | - | 2 - 3 | 120 | 3156501 | 137 | |
BC012308.1 | - | 2 - 3 | 147 | 3500167 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |