Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013867.2 | NP_038895.1 | 7 - 8 | 2085 | 77 | |
GenBank mRNA | AF179566.1 | - | 7 - 8 | 2033 | 77 | |
AK143894.1 | - | 7 - 8 | 1873 | 77 | ||
AK158241.1 | - | 7 - 8 | 2086 | 77 | ||
AK167578.1 | - | 7 - 8 | 1904 | 77 | ||
BC023930.1 | - | 7 - 8 | 2074 | 5324365 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |