Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011836.3 | NP_035966.2 | 15 - 16 | 2846 | 80 | |
GenBank mRNA | AF079520.1 | - | 15 - 16 | 2836 | 80 | |
AF083372.1 | - | 15 - 16 | 2783 | 80 | ||
AK132948.1 | - | 12 - 13 | 1831 | 80 | ||
BC096366.1 | - | 15 - 16 | 2846 | 2865309 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |