Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081083.2 | NP_001074552.1 | 2 - 3 | 288 | 101 | |
NM_001271563.1 | NP_001258492.1 | 2 - 3 | 288 | 101 | ||
NM_001271564.1 | NP_001258493.1 | 2 - 3 | 288 | 101 | ||
NM_001271565.1 | NP_001258494.1 | 2 - 3 | 288 | 101 | ||
GenBank mRNA | AK014880.1 | - | 2 - 3 | 105 | 101 | |
AK041744.1 | - | 2 - 3 | 112 | 101 | ||
BC145004.1 | - | 2 - 3 | 96 | 101 | ||
BC145005.1 | - | 2 - 3 | 96 | 9053553 | 101 | |
BC145013.1 | - | 2 - 3 | 96 | 9053561 | 101 | |
BC150688.1 | - | 2 - 3 | 96 | 9087599 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |