Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164274.1 | NP_001157746.1 | 13 - 14 | 4594 | 79 | |
NM_013720.2 | NP_038748.2 | 13 - 14 | 4597 | 79 | ||
GenBank mRNA | AF205935.1 | - | 12 - 13 | 4414 | 79 | |
AK172962.1 | - | 13 - 14 | 4517 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |