Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164274.1 | NP_001157746.1 | 7 - 8 | 2603 | 106 | |
NM_013720.2 | NP_038748.2 | 7 - 8 | 2603 | 106 | ||
GenBank mRNA | AK046252.1 | - | 7 - 8 | 2604 | 106 | |
AK139919.1 | - | 7 - 8 | 2556 | 106 | ||
AK172962.1 | - | 7 - 8 | 2526 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |