Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nusap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042652.1 | NP_001036117.1 | 7 - 8 | 1364 | 75 | |
NM_133851.3 | NP_598612.1 | 8 - 9 | 1463 | 75 | ||
GenBank mRNA | AF305710.1 | - | 8 - 9 | 1044 | 75 | |
AK042697.1 | - | 945 | 75 | |||
AK044819.1 | - | 8 - 9 | 1036 | 75 | ||
AK076028.1 | - | 8 - 9 | 1033 | 75 | ||
AK088389.1 | - | 8 - 9 | 1063 | 75 | ||
BC004787.1 | - | 8 - 9 | 1013 | 3588697 | 75 | |
BC100392.1 | - | 8 - 9 | 1463 | 30110191 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |