Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021526.2 | NP_067501.2 | 7 - 8 | 781 | 103 | |
GenBank mRNA | AK011974.1 | - | 7 - 8 | 730 | 103 | |
AK012013.1 | - | 7 - 8 | 712 | 103 | ||
AK150280.1 | - | 7 - 8 | 717 | 103 | ||
AK151104.1 | - | 7 - 8 | 764 | 103 | ||
AK151733.1 | - | 7 - 8 | 764 | 103 | ||
BC003742.1 | - | 7 - 8 | 686 | 3599671 | 103 | |
Y13071.1 | - | 7 - 8 | 702 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |