Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271467.1 | NP_001258396.1 | 5 - 6 | 801 | 122 | |
NM_001271468.1 | NP_001258397.1 | 4 - 5 | 703 | 122 | ||
NM_001271470.1 | NP_001258399.1 | 5 - 6 | 868 | 122 | ||
NM_001271492.1 | NP_001258421.1 | 4 - 5 | 703 | 122 | ||
NM_010249.2 | NP_034379.1 | 4 - 5 | 703 | 122 | ||
NM_207669.2 | NP_997552.1 | 4 - 5 | 703 | 122 | ||
GenBank mRNA | AK040722.1 | - | 4 - 5 | 704 | 122 | |
AK052828.1 | - | 679 | 122 | |||
AK139710.1 | - | 5 - 6 | 868 | 122 | ||
AK140222.1 | - | 5 - 6 | 754 | 122 | ||
AK140907.1 | - | 4 - 5 | 614 | 122 | ||
AK146373.1 | - | 4 - 5 | 567 | 122 | ||
AY282802.1 | - | 2 - 3 | 355 | 122 | ||
BC013558.1 | - | 4 - 5 | 594 | 3592119 | 122 | |
CA552206.1 | - | 1 - 2 | 120 | 122 | ||
CF730595.1 | - | 4 - 5 | 556 | 122 | ||
M74516.1 | - | 4 - 5 | 598 | 122 | ||
M74517.1 | - | 4 - 5 | 597 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |