Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010725.2 | NP_034855.2 | 3 - 4 | 488 | 92 | |
GenBank mRNA | AB221651.1 | - | 3 - 4 | 488 | 92 | |
AF078166.1 | - | 3 - 4 | 488 | 92 | ||
BC119169.1 | - | 3 - 4 | 567 | 8733918 | 92 | |
BC125469.1 | - | 3 - 4 | 567 | 40129984 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |