Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jarid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205043.1 | NP_001191972.1 | 16 - 17 | 4041 | 71 | |
NM_001205044.1 | NP_001191973.1 | 17 - 18 | 3714 | 71 | ||
NM_021878.3 | NP_068678.1 | 17 - 18 | 3760 | 71 | ||
GenBank mRNA | AK045214.1 | - | 16 - 17 | 3846 | 71 | |
AK147226.1 | - | 16 - 17 | 4041 | 71 | ||
BC003374.1 | - | 10 - 11 | 1610 | 71 | ||
BC026056.1 | - | 16 - 17 | 3948 | 71 | ||
BC050129.1 | - | 16 - 17 | 3962 | 71 | ||
BC051010.1 | - | 14 - 15 | 6402 | 71 | ||
BC052444.1 | - | 17 - 18 | 3631 | 6406875 | 71 | |
BC060695.1 | - | 17 - 18 | 3631 | 6406875 | 71 | |
BC115758.1 | - | 11 - 12 | 2645 | 71 | ||
D31967.1 | - | 17 - 18 | 3650 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |