Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031389.2 | NP_113566.2 | 2 - 3 | 405 | 75 | |
GenBank mRNA | AK133284.1 | - | 2 - 3 | 406 | 75 | |
AK166817.1 | - | 2 - 3 | 386 | 75 | ||
AK166869.1 | - | 2 - 3 | 631 | 75 | ||
AY596194.1 | - | 2 - 3 | 405 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |