Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145838.1 | NP_665837.1 | 3 - 4 | 363 | 127 | |
GenBank mRNA | AB059554.1 | - | 3 - 4 | 363 | 127 | |
AK085105.1 | - | 4 - 5 | 500 | 127 | ||
AK156557.1 | - | 3 - 4 | 900 | 127 | ||
AK172400.1 | - | 3 - 4 | 428 | 127 | ||
BC115912.1 | - | 3 - 4 | 318 | 40104779 | 127 | |
BC118063.1 | - | 3 - 4 | 318 | 40104780 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |