Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023668.2 | NP_076157.2 | 3 - 4 | 441 | 82 | |
GenBank mRNA | AF290472.1 | - | 3 - 4 | 373 | 82 | |
AF323918.1 | - | 3 - 4 | 461 | 82 | ||
AK011168.1 | - | 3 - 4 | 351 | 82 | ||
AK146029.1 | - | 3 - 4 | 370 | 82 | ||
AK212133.1 | - | 2 - 3 | 220 | 82 | ||
BC021434.1 | - | 337 | 5029832 | 82 | ||
BC046796.1 | - | 3 - 4 | 372 | 5718032 | 82 | |
CJ092278.1 | - | 3 - 4 | 442 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |