Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012305.2 | NP_001012305.1 | 10 - 11 | 1775 | 77 | |
GenBank mRNA | BC089362.1 | - | 10 - 11 | 1769 | 30628112 | 77 |
BC113764.1 | - | 10 - 11 | 1758 | 40059402 | 77 | |
BC113765.1 | - | 10 - 11 | 1758 | 40059404 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |