Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009661.4 | NP_033791.1 | 10 - 11 | 1490 | 87 | |
GenBank mRNA | AK028724.1 | - | 10 - 11 | 1486 | 87 | |
AK158553.1 | - | 8 - 9 | 1364 | 87 | ||
BC015253.1 | - | 10 - 11 | 1490 | 4221815 | 87 | |
U93277.1 | - | 10 - 11 | 1456 | 87 | ||
Y14696.1 | - | 10 - 11 | 1469 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |