Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013256.2 | NP_001013274.2 | 15 - 16 | 2874 | 70 | |
NM_001143790.1 | NP_001137262.1 | 15 - 16 | 2871 | 70 | ||
NM_001281464.1 | NP_001268393.1 | 15 - 16 | 2742 | 70 | ||
NM_001281465.1 | NP_001268394.1 | 15 - 16 | 2858 | 70 | ||
GenBank mRNA | AK044055.1 | - | 15 - 16 | 2653 | 70 | |
AK145919.1 | - | 15 - 16 | 2689 | 70 | ||
AK165906.1 | - | 15 - 16 | 2674 | 70 | ||
AK197949.1 | - | 318 | 70 | |||
AK198189.1 | - | 1 - 2 | 147 | 70 | ||
AK213842.1 | - | 3 - 4 | 322 | 70 | ||
BC014687.1 | - | 6 - 7 | 1042 | 70 | ||
BC060629.1 | - | 11 - 12 | 1924 | 6847619 | 70 | |
BC066078.1 | - | 15 - 16 | 2685 | 70 | ||
BC072625.1 | - | 15 - 16 | 2699 | 70 | ||
BC147495.1 | - | 15 - 16 | 2672 | 9056325 | 70 | |
BC147497.1 | - | 15 - 16 | 2672 | 9056354 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |