Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100007294.mm.1 |
NM_177779.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007294.mm.1 |
NM_177779.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007294.mm.1 |
TC1100000876.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007292.mm.1 |
NM_177779.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007292.mm.1 |
NM_177779.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007292.mm.1 |
TC1100000876.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007293.mm.1 |
NM_177779.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007293.mm.1 |
NM_177779.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007293.mm.1 |
TC1100000876.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100007295.mm.1 |
NM_177779.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007295.mm.1 |
NM_177779.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100007295.mm.1 |
TC1100000876.mm.1 |
MTA TC |