Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rreb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039188.1 | NP_001034277.1 | 6 - 7 | 1009 | 78 | |
NM_001177868.1 | NP_001171339.1 | 6 - 7 | 734 | 78 | ||
NM_001177869.1 | NP_001171340.1 | 5 - 6 | 670 | 78 | ||
NM_026830.2 | NP_081106.1 | 6 - 7 | 977 | 78 | ||
NR_033218.1 | 6 - 7 | 2131 | 78 | |||
NR_033615.1 | 6 - 7 | 1948 | 78 | |||
GenBank mRNA | AK047047.1 | - | 6 - 7 | 1928 | 78 | |
AK047251.1 | - | 6 - 7 | 1930 | 78 | ||
AK147260.1 | - | 6 - 7 | 2132 | 78 | ||
AK147653.1 | - | 6 - 7 | 2132 | 78 | ||
AK154980.1 | - | 6 - 7 | 977 | 78 | ||
AK170652.1 | - | 6 - 7 | 1009 | 78 | ||
AK171375.1 | - | 6 - 7 | 714 | 78 | ||
AY946044.1 | - | 3 - 4 | 361 | 78 | ||
BC080680.1 | - | 6 - 7 | 823 | 6330949 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |