Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170779.1 | NP_740749.1 | 18 - 19 | 2631 | 99 | |
GenBank mRNA | AB271907.1 | - | 18 - 19 | 2601 | 99 | |
AK220259.1 | - | 3002 | 99 | |||
BC006733.1 | - | 15 - 16 | 2220 | 99 | ||
BC017638.1 | - | 11 - 12 | 1757 | 99 | ||
BC037006.1 | - | 10 - 11 | 1479 | 5116674 | 99 | |
DQ256090.1 | - | 18 - 19 | 2631 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |