Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aurkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080965.1 | NP_001074434.1 | 3 - 4 | 394 | 153 | |
NM_001080966.1 | NP_001074435.1 | 3 - 4 | 680 | 153 | ||
NM_020572.2 | NP_065597.2 | 3 - 4 | 558 | 153 | ||
GenBank mRNA | AF054620.1 | - | 3 - 4 | 645 | 153 | |
BC064780.1 | - | 3 - 4 | 562 | 30476803 | 153 | |
BC100337.1 | - | 3 - 4 | 315 | 153 | ||
BG080448.2 | - | 3 - 4 | 371 | 153 | ||
DV648982.1 | - | 3 - 4 | 422 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |